Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLH1B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987318
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7318 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7318 Supplier Bioss Supplier No. BS6535R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PSMAL has both folate hydrolase and N-acetylated-alpha-linked-acidic dipeptidase (NAALADase) activity. PSMAL exhibits a dipeptidyl-peptidase IV type activity.

Specifications

Antigen FOLH1B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FOLH1B
Gene Accession No. Q9HBA9
Gene Alias Cell growth-inhibiting gene 26 protein; folate hydrolase 1B; folate hydrolase 2; folate hydrolase pseudogene; FOLH1B; FOLH2; FOLHP; GCP3; GCPIII; GIG26; glutamate carboxypeptidase III; NAALADase; N-acetylated-alpha-linked-acidic dipeptidase; Prostate-specific membrane antigen-like protein; PSM; PSMAL; PSMA-LIKE; Putative folate hydrolase 1B; Putative N-acetylated-alpha-linked acidic dipeptidase
Gene Symbols FOLH1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FOLH1B .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219595
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.