Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Folic Acid, MP Biomedical

Catalog No. MP210172525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210172525 25 g
MP210172510 10 g
MP210172580 100 g
MP210172505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP210172525 Supplier MP Biomedicals, Inc Supplier No. 0210172525
Add to Cart
Add to Cart

Folic acid >95% (Vitamin B9 and folate) is essential to numerous bodily functions. The human body needs folate to synthesize DNA, repair DNA, and methylate DNA as well as to act as a cofactor in certain biological reactions. It is especially important in aiding rapid cell division and growth, such as in infancy and pregnancy.

  • Folic acid can be found in a variety of fruits and vegetables
  • Water-soluble vitamin
  • Helps the body form red blood cells and aids in the formation of genetic material within every body cell
  • Exhibits metal binding properties
  • Hematopoietic vitamin present, free or combined with one or more additional molecules of L-(+)-glutamic acid, in liver, kidney, mushrooms, spinach, yeast, green leaves, and grasses
  • Folic acid (FA) and dihydrofolic acid (FAH2) are substrates of dihydrofolate reductase(s) which reduce them to tetrahydrofolate (THF), which in turn supports ‘one carbon’ transfer
  • Tetrahydrofolates are required for de novo synthesis of purines, thymidylic acid, and various amino acids and for post-translational methylation (epigenetics)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.40
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
PubChem CID 6037
ChEBI CHEBI:27470

Specifications

Color Orange
pH 4.0 to 4.8
Packaging Glass
Quantity 25 g
Percent Purity 95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.