Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Folin-Ciocalteu Phenol TS Test Solution, USP, Spectrum™ Chemical
SDP

Catalog No. 18602809 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle

F-184

Spectrum™ Chemical Folin-Ciocalteu Phenol TS, USP Test Solution is a solution, sometimes designated with 'TS or Test Solution', that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.

Chemical Identifiers

CAS 7732-18-5, 10377-48-7, 7664-38-2, 7647-01-0, 10213-10-2, 10102-40-6
Molecular Formula H4Na2O6W
Molecular Weight (g/mol) 329.85
InChI Key WPZFLQRLSGVIAA-UHFFFAOYSA-N
IUPAC Name disodium dioxotungstenbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O

Specifications

CAS Max % 80%, <10%, <5%, <5%, <3%, <3%
UN Number UN3264
Grade Chemical Solution
Packaging Amber Glass Bottle
pH Acidic
Quantity 500 mL
Chemical Name or Material Folin-Ciocalteu Phenol TS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.