Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Folinic acid calcium salt hydrate, ≥90%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
100 mg
500 mg
1 g
Description
Featured Industries: Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 1492-18-8 (anhydrous) |
| Physical Form | Neat |
| Linear Formula | C20H21 CaN7O7 · xH2 O |
| Melting Point | 240°C to 250°C |
| Synonym | 5-Formyl-5,6,7,8-tetrahydrofolic acid calcium salt; 5-Formyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid calcium salt; 5-HCO-H4 PteGlu; Calcium folinate; Citrovorum factor calcium salt; Leucovorin calcium salt |
| Type | Analytical Standard |
| Quantity | 1 g |
| Formula Weight | 511.50 (anhydrous basis) |
| Percent Purity | ≥90% |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction