Learn More
Invitrogen™ Follicle Stimulating Hormone Monoclonal Antibody (P2H3)

Description
MIF2710 targets Follicle Stimulating Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MIF2710 immunogen is human Follicle Stimulating Hormone. MIF2710 detects Follicle Stimulating Hormone which has a predicted molecular weight of approximately 13 kDa. MIF2710 was formerly sold as a Seradyn product.
Specifications
Specifications
| Antigen | Follicle Stimulating Hormone |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | P2H3 |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | FSHB |
| Gene Accession No. | P01215, P01225 |
| Gene Alias | class 1 follicle-stimulating hormone beta polypeptide; class 2 follicle-stimulating hormone beta polypeptide; follicle - stimulating hormone subunit beta; follicle stimulating hormone beta; follicle stimulating hormone beta subunit; follicle stimulating hormone beta-subunit precursor; follicle stimulating hormone subunit beta; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; follicle-stimulating hormone beta-subunit; follicle-stimulating hormone subunit beta; follicule stimulating hormone, beta polypeptide; follitropin beta chain; Follitropin subunit beta; follitropin, beta chain; FSH; FSH-alpha; FSHB; FSH-B; Fshbeta; FSH-beta; HH24; LSH-alpha; prefollicle stimulating hormone beta chain; TSH-alpha |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.