Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FSH beta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579283
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579283 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579283 Supplier Invitrogen™ Supplier No. PA579283
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - IHC: mouse brain tissue, human mammry cancer tissue.

Human follicle stimulating hormone is a glycoprotein produced from urine of post-menopausal women and having a total molecular mass of 35,500 Dalton. HuFSH is a heterodimeric hormone consisting of 165 amino acids a chain (non-active) and 115 amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FSH beta
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene FSHB
Gene Accession No. P01225, Q60687
Gene Alias follicle stimulating hormone beta subunit; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; Follitropin beta chain; follitropin subunit beta; follitropin, beta chain; FSHB; FSH-B; FSH-beta; HH24
Gene Symbols FSHB
Host Species Rabbit
Immunogen E.coli-derived human FSH beta recombinant protein (Position: N19-E129).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14308, 2488
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.