Learn More
Invitrogen™ Follicle Stimulating Hormone Recombinant Rabbit Monoclonal Antibody (9M9C5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA558173

Description
Immunogen sequence: APDVQDCPEC TLQENPFFSQ PGAPILQCMG CCFSRAYPTP LRSKKTMLVQ KNVTSESTCC VAKSYNRVTV MGGFKVENHT ACHCSTCYYH KS.
Follicle-stimulatine hormone (FSH) is a member of the glycoprotein hormone family. Members of this family consist of an identical alpha subunit and a hormone-specific beta subunit. Alternative splicing results in two transcript variants encoding the same protein. FSH is secreted by gonadotropic cells in the pituitary gland and in conjunction with luteinizing hormone, induces egg and sperm production.
Specifications
Follicle Stimulating Hormone | |
Recombinant Monoclonal | |
0.57 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P01225 | |
FSHB | |
Recombinant fusion protein containing a sequence corresponding to amino acids 25-116 of human FSH alpha/beta (NP_000726.1/NP_000501.1). | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunocytochemistry | |
9M9C5 | |
Unconjugated | |
FSHB | |
class 1 follicle-stimulating hormone beta polypeptide; class 2 follicle-stimulating hormone beta polypeptide; follicle - stimulating hormone subunit beta; follicle stimulating hormone beta; follicle stimulating hormone beta subunit; follicle stimulating hormone beta-subunit precursor; follicle stimulating hormone subunit beta; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; follicle-stimulating hormone beta-subunit; follicle-stimulating hormone subunit beta; follicule stimulating hormone, beta polypeptide; follitropin beta chain; Follitropin subunit beta; follitropin, beta chain; FSH; FSH-alpha; FSHB; FSH-B; Fshbeta; FSH-beta; HH24; LSH-alpha; prefollicle stimulating hormone beta chain; TSH-alpha | |
Rabbit | |
Protein A | |
RUO | |
2488 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.