Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Folliculin Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$207.00 - $499.50

Specifications

Antigen Folliculin
Dilution Western Blot 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50-1:200
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP144995SS
SDP
View Documents
Novus Biologicals
NBP144995SS
0.025 mL
Each for $207.00
Only null left
Add to Cart
 
NBP144995
SDP
View Documents
Novus Biologicals
NBP144995
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Folliculin Polyclonal specifically detects Folliculin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Folliculin
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cell Cycle and Replication
Whole antisera with 0.1% Sodium Azide
BHD skin lesion fibrofolliculoma protein, BHDFLCL, Birt-Hogg-Dube syndrome protein, DKFZp547A118, FLJ45004, FLJ99377, folliculin, MGC17998, MGC23445
FLCN
IgG
Unpurified
This antibody is expected to detect all three isoforms of Folliculin protein ( UniProt # Q8NFG4)
Western Blot 1:1000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50-1:200
Polyclonal
Antisera
RUO
Human, Mouse
Q8NFG4, Q8QZS3
201163
N-terminal partial recombinant human Folliculin protein [Swiss-Prot# Q8NFG4] expressed in E. coli.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.