Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Folliculin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120278
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120278 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120278 Supplier Invitrogen™ Supplier No. PA5120278
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse brain, Mouse thymus, Rat heart. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: RKLPVFKSLR HMRQVLGAPS FRMLAWHVLM GNQVIWKSRD VDLVQSAFEV LRTMLPVGCV RIIPYSSQYE EAYRCNFLGL SPHVQIPPHV LSSEFAVIVE VHAAARSTLH PVGCEDDQSL SKYEFVVTSG SPVAADRVGP TILNKIEAAL TNQNLSVDVV DQCLVCLKEE WMNKVKVLFK FTKVDSRPKE DTQKLLSILG ASEEDNVKLL KFWMTGLSKT YKSHLMSTVR SPTASESRN.

This gene encodes a mitochondrial protein that is characterized by a poly-proline rich region. A chicken homolog of this protein promotes mitochondrial fission and the mouse homolog protects cells from oxidative stress. A related pseudogene of this gene is found on chromosome X.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Folliculin
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FLCN
Gene Accession No. Q76JQ2, Q8NFG4, Q8QZS3
Gene Alias AU014660; B430214A04Rik; BHD; BHD skin lesion fibrofolliculoma protein; birt-Hogg-Dube syndrome protein; Birt-Hogg-Dube syndrome protein homolog; FLCL; Flcn; Folliculin
Gene Symbols FLCN
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 341-579 of human FLCN (NP_659434.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201163, 216805, 303185
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.