Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Follistatin Antibody (FST/4282), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308814AF350
Description
Follistatin Monoclonal antibody specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, ImmunocytochemistrySpecifications
Follistatin | |
Monoclonal | |
Alexa Fluor 350 | |
follistatin, follistatin isoform FST317, FSActivin-binding protein | |
Recognizes human Follistatin isoform FS288, raised against recombinant human FS288. (Uniprot: P19883) | |
100 μg | |
Cancer | |
10468 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry | |
FST/4282 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction