Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Follistatin Antibody (H10), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261940AF488
Description
Follistatin Monoclonal specifically detects Follistatin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Follistatin | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FST | |
Synthetic peptide corresponding to sequence CDEDQDYSFPISSILEW of the C- terminal region of human Follistatin 315 | |
0.1 mL | |
Primary | |
This antibody recognizes human Follistatin isoform FS315. | |
Store at 4°C in the dark. | |
IgG2a |
ELISA, Immunohistochemistry, Western Blot | |
H10 | |
Western Blot, ELISA, Immunohistochemistry | |
follistatin, follistatin isoform FST317, FSActivin-binding protein | |
Mouse | |
Protein A purified | |
Cancer | |
10468 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction