Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Follistatin Monoclonal Antibody (C13), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA551035
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551035 200 μL
FEMA551036 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551035 Supplier Invitrogen™ Supplier No. MA551035
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Follistatin is a single-chain gonadal protein that specifically inhibits follicle-stimulating hormone release. The single FST gene encodes two isoforms, FST317 and FST344 containing 317 and 344 amino acids respectively, resulting from alternative splicing of the precursor mRNA. In a study in which 37 candidate genes were tested for linkage and association with polycystic ovary syndrome (PCOS) or hyperandrogenemia in 150 families, evidence was found for linkage between PCOS and follistatin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Follistatin
Applications Western Blot
Classification Monoclonal
Clone C13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Fst
Gene Accession No. P19883, P21674
Gene Alias activin-binding protein; AL033346; FOL1; Follictatin; Follistatin; follistatin isoform FST317; follistatin-A; FS; FST; Fst-288; RATFOL1
Gene Symbols Fst
Host Species Mouse
Immunogen Recombinant Human FS protein, Arg35-Val283 (Accession #P19883).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10468, 24373
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.