Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOLR1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169363
Description
FOLR1 Polyclonal specifically detects FOLR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
FOLR1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, FOLR, FR-alpha, KB cells FBP, Ovarian tumor-associated antigen MOv18 | |
Rabbit | |
29 kDa | |
100 μL | |
Cancer | |
2348 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
P15328 | |
FOLR1 | |
Synthetic peptides corresponding to FOLR1(folate receptor 1 (adult)) The peptide sequence was selected from the middle region of FOLR1. Peptide sequence HFYFPTPTVLCNEIWTHSYKVSNYSRGSGRCIQMWFDPAQGNPNEEVARF. | |
Affinity purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction