Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR1 Mouse anti-Human, Clone:2G5C12, Abnova™

Catalog No. 89333932 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-932 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-932 Supplier Abnova Corporation Supplier No. MAB17630
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human FOLR1.

The protein encoded by this gene is a member of the folate receptor (FOLR) family. Members of this gene family have a high affinity for folic acid and for several reduced folic acid derivatives, and mediate delivery of 5-methyltetrahydrofolate to the interior of cells. This gene is composed of 7 exons; exons 1 through 4 encode the 5′ UTR and exons 4 through 7 encode the open reading frame. Due to the presence of 2 promoters, multiple transcription start sites, and alternative splicing of exons, several transcript variants are derived from this gene. These variants differ in the lengths of 5′ and 3′ UTR, but they encode an identical amino acid sequence. [provided by RefSeq]

Specifications

Antigen FOLR1
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2G5C12
Conjugate Unconjugated
Description folate receptor 1 (adult)
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunocytochemistry (1:100-1:500)
Flow Cytometry (1:200-1:400)
Immunohistochemistry (1:200-1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene FOLR1
Gene Alias FBP, FOLR, FR-alpha, MOv18
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 41-227 of human FOLR1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 2348
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.