Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOLR2 Antibody (046), DyLight 405 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289847V
Description
FOLR2 Monoclonal antibody specifically detects FOLR2 in Human samples. It is validated for ELISASpecifications
FOLR2 | |
Monoclonal | |
DyLight 405 | |
BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human FOLR2 (P14207-1; Met 1-His 228). | |
0.1 mL | |
Cancer | |
2350 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
046 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction