Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOLR2 Antibody (CL10), FITC, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP281079F
Description
FOLR2 Monoclonal specifically detects FOLR2 in Mouse samples. It is validated for Immunohistochemistry.Specifications
FOLR2 | |
Monoclonal | |
FITC | |
PBS | |
FOLR2 | |
FRbeta -transfected RBL2H3 cells. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry | |
CL10 | |
Immunohistochemistry | |
BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein | |
Rat | |
Protein A purified | |
RUO | |
2350 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction