Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOLR2 Monoclonal Antibody (1905CT501.41.76)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA537763
Description
FOLR2 Monoclonal Antibody for Western Blot
FOLR2 is a member of the folate receptor (FOLR) family, and these genes exist in a cluster on chromosome 11. Members of this gene family have a high affinity for folic acid and for several reduced folic acid derivatives, and they mediate delivery of 5-methyltetrahydrofolate to the interior of cells. This protein has a 68% and 79% sequence homology with the FOLR1 and FOLR3 proteins, respectively. Although this protein was originally thought to be specific to placenta, it can also exist in other tissues, and it may play a role in the transport of methotrexate in synovial macrophages in rheumatoid arthritis patients.
Specifications
FOLR2 | |
Monoclonal | |
0.25 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P14207 | |
FOLR2 | |
recombiant protein from human FOLR2. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Western Blot | |
1905CT501.41.76 | |
Unconjugated | |
FOLR2 | |
BETA-HFR; FBP; FBP/PL-1; Fbp2; folate binding protein; folate binding protein 2; folate receptor 2; folate receptor 2 (fetal); folate receptor beta; folate receptor, beta; folate receptor, fetal/placental; Folate-binding protein 2; folate-binding protein, fetal/placental; Folbp2; Folbp-2; FOLR2; FR-beta; FR-P3; placental folate-binding protein | |
Mouse | |
Protein G | |
RUO | |
2350 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction