Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOLR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547764
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547764 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547764 Supplier Invitrogen™ Supplier No. PA547764
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) FOLR1 and rhFOLR3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

FOLR2 is a member of the folate receptor (FOLR) family, and these genes exist in a cluster on chromosome 11. Members of this gene family have a high affinity for folic acid and for several reduced folic acid derivatives, and they mediate delivery of 5-methyltetrahydrofolate to the interior of cells. This protein has a 68% and 79% sequence homology with the FOLR1 and FOLR3 proteins, respectively. Although this protein was originally thought to be specific to placenta, it can also exist in other tissues, and it may play a role in the transport of methotrexate in synovial macrophages in rheumatoid arthritis patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOLR2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene FOLR2
Gene Accession No. P14207
Gene Alias BETA-HFR; FBP; FBP/PL-1; Fbp2; folate binding protein; folate binding protein 2; folate receptor 2; folate receptor 2 (fetal); folate receptor beta; folate receptor, beta; folate receptor, fetal/placental; Folate-binding protein 2; folate-binding protein, fetal/placental; Folbp2; Folbp-2; FOLR2; FR-beta; FR-P3; placental folate-binding protein
Gene Symbols FOLR2
Host Species Sheep
Immunogen CHO-derived recombinant human FOLR2 Gln22-His228.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2350
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.