Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOLR2, Rat anti-Mouse, Clone: CL10, Novus Biologicals™
SDP

Catalog No. p-7182734 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015546 0.025 mg
NB015547 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB015546 Supplier Novus Biologicals Supplier No. NBP2810790.025MG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

FOLR2 Monoclonal specifically detects FOLR2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FOLR2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone CL10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias BETA-HFR, FBP, FBP/PL-1, Folate receptor 2, folate receptor 2 (fetal), folate receptor beta, folate receptor, beta, Folate receptor, fetal/placental, folate-binding protein, fetal/placental, FR-BETA, FR-P3, Placental folate-binding protein
Gene Symbols FOLR2
Host Species Rat
Immunogen FRbeta -transfected RBL2H3 cells.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2350
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.