Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOLR4 Antibody (12A5), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP226652MFV610
Description
FOLR4 Monoclonal antibody specifically detects FOLR4 in Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
FOLR4 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
390243 | |
Store at 4C in the dark. | |
IgG2b κ |
Flow Cytometry | |
12A5 | |
Flow Cytometry, Flow (Intracellular) | |
folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta | |
CD3 mAb-stimulated CD4/CD25 T cells | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction