Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOLR4 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299508F
Description
FOLR4 Polyclonal antibody specifically detects FOLR4 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
FOLR4 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
folate receptor 4 (delta) homolog (mouse), Folbp3, probable folate receptor delta | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FOLR4. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
390243 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction