Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOLR4 Monoclonal Antibody (12A5)

Catalog No. PIMA524803
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524803 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524803 Supplier Invitrogen™ Supplier No. MA524803

Rat Monoclonal Antibody

FOLR4 Monoclonal Antibody for Flow

FR4 is a heavily glycosylated 35 kD receptor for folic acid and the physiologic circulating form of the vitamin, N5-methyltetrahydrofolate. Natural T regs have high levels of FR4 and together with CD25 can be used to distinguish natural Tregs, effector T cells, memory-like T cells and naive T cells. In vivo studies with eBioTH6 show a decrease in the number of CD4+CD25+ cells and an enhanced tumor response leading to the elimination of advanced stages of tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOLR4
Applications Flow Cytometry
Classification Monoclonal
Clone 12A5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene Izumo1r
Gene Accession No. Q9EQF4
Gene Alias 0910001L11Rik; C86255; Folate receptor 4; folate receptor 4 (delta); folate receptor 4 (delta) homolog; folate receptor 4, delta (putative); folate receptor delta; Folate-binding protein 3; Folbp3; Folr4; FR-delta; IZUMO1 receptor protein JUNO; IZUMO1 receptor, JUNO; Izumo1r; Juno; probable folate receptor delta; sperm-egg fusion protein Juno
Gene Symbols Izumo1r
Host Species Rat
Immunogen CD3 mAb-stimulated CD4/CD25 T cells.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64931
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.