Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOLR4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544954
Description
Peptide sequence: TCKSNWRGGW DWSQGKNRCP KGAQCLPFSH YFPTPADLCE KTWSNSFKAS Sequence homology: Guinea Pig: 85%; Horse: 90%; Human: 100%.
FR4 is a heavily glycosylated 35 kD receptor for folic acid and the physiologic circulating form of the vitamin, N5-methyltetrahydrofolate. Natural T regs have high levels of FR4 and together with CD25 can be used to distinguish natural Tregs, effector T cells, memory-like T cells and naive T cells. In vivo studies with eBioTH6 show a decrease in the number of CD4+CD25+ cells and an enhanced tumor response leading to the elimination of advanced stages of tumors.
Specifications
| FOLR4 | |
| Polyclonal | |
| Unconjugated | |
| Izumo1r | |
| 0910001L11Rik; C86255; Folate receptor 4; folate receptor 4 (delta); folate receptor 4 (delta) homolog; folate receptor 4, delta (putative); folate receptor delta; Folate-binding protein 3; Folbp3; Folr4; FR-delta; IZUMO1 receptor protein JUNO; IZUMO1 receptor, JUNO; Izumo1r; Juno; probable folate receptor delta; sperm-egg fusion protein Juno | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 390243 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A6ND01 | |
| Izumo1r | |
| Synthetic peptide directed towards the middle region of human FOLR4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction