Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOPNL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559216 Supplier Invitrogen™ Supplier No. PA559216
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IHELNAFEES KDNTIPLLYG ILAHFLRGTK DGIQNAFLKG PSLQPSDPSL GRQPSRRKPM DDHLRKEEQK STNIEDLHVS Q Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 58%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FOPNL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FOPNL
Gene Accession No. Q96NB1
Gene Alias 0610037P05Rik; C16orf63; FGFR1OP N-terminal like; FGFR1OP N-terminal-like protein; FOPNL; FOP-related protein of 20 kDa; FOR20; lisH domain-containing protein C16orf63; lisH domain-containing protein C16orf63 homolog; lisH domain-containing protein FOPNL; PHSECRG2; pluripotent embryonic stem cell-related protein; RGD1305823
Gene Symbols FOPNL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FOPNL. Recombinant protein control fragment (Product #RP-98203).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123811
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.