Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Forceps, 4 Inch - Blunt End - Stainless Steel
SCI_ED

Catalog No. S02943 Shop All Eisco Products
Change view
Click to view available options
Length (Metric):
10 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S02943 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02943 Supplier Eisco™ Supplier No. BI0170A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eisco Labs 4 in. forceps with blunt ends. Made of stainless steel for durability. Ideal for precise handling and manipulation in laboratory and educational settings.

  • Stainless steel, 10 mm

Specifications

Length (English) 4 in.
Length (Metric) 10 cm
Material Stainless Steel
Style Standard
Tip Style Blunt
Product Type Tweezers/Forceps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.