Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensic Chemistry: Chemical Detection of Fingerprints
SCI_ED

Catalog No. s96357
Click to view available options
Age:
13+ years
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.
WARNING: This product can expose you to chemicals including Methanol/ Methyl Isobutyl Ketone, which is known to the State of California to cause cancer and reproductive harm.

Includes

Teacher's manual and student guide, analysis copy-masters, one tube extra bonding glue, one bottle ninhydrin powder, four bottles 95% ethanol, 15 fuming trays, 15 black plastic sheets

Three activities include fingerprint analysis, ninhydrin development and cyanoacrylate fuming.

  • Use alternative methods for detecting fingerprints
  • Examine other methods that are used for gathering evidence when dusting for fingerprints is not effective
  • Learn to identify fingerprint types, a method of fuming for fingerprints and a technique of chemically developing fingerprints
  • Class size: 15 students

Specifications

Certifications/Compliance 49 CFR 173.4
Product Type Chemical Detection of Fingerprints Kit
Color White
Grade 7 to 14
Age 13+ years
For Use With (Application) Detecting fingerprints
Includes Teachers manual and student guide, analysis copy-masters, one tube extra bonding glue, one bottle ninhydrin powder, four bottles 95% ethanol, 15 fuming trays, 15 black plastic sheets
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Students
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.