Learn More
Description
Includes
Teacher's manual and student guide, analysis copy-masters, fingerprinting powder, 15 fingerprinting brushes, 15 hand magnifiers, ink pad and acetate sheets. Enough materials for 15 tests.
- Investigate the oldest and most commonly used method of fingerprint detection — dusting for fingerprints
- Learn how to identify different types of fingerprints and distinguishing characteristics
Specifications
Specifications
| Certifications/Compliance | 49 CFR 173.4 |
| Color | White |
| Grade | 7 to 14 |
| Age | 13 to 20 |
| For Use With (Application) | Fingerprint analysis |
| Includes | 1 bottle fingerprinting powder, 15 fingerprinting brushes, 15 hand magnifiers, ink pad, acetate sheets, Teacher's manual and student study guide copymasters |
| Quantity | 1 |
| Dimensions (L x W x H) | 9.75 x 6.75 x 3 in. |
| Class Size | 15 Lab Groups |
| Product Type | Dusting For Fingerprints Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

