Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Forma™ Freezer Replacement Shelves
Click to view available options
For Use With (Equipment):
17.3 cu. ft. Freezers
Type:
Shelf Kit
Description
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
- Stainless steel
- Kits include mounting hardware
Specifications
Specifications
| Type | Shelf Kit |
| Capacity (English) | 17.3 cu. ft. |
| Capacity (Metric) | 489.8 L |
| Style | Upright |
| Color | Silver |
| For Use With (Equipment) | 17.3 cu. ft. Freezers |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction