Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formalde-Fresh Solution, Buffered, Fisher Chemical™

Catalog No. p-21735 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle
PolyPac™
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SF93-4 4 L Poly Bottle
SF93-20 20 L PolyPac™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SF93-4
Only null left
Add to Cart
This item is not returnable. View return policy

Specifications

Physical Form Liquid
Chemical Name or Material Formalde-Fresh Solution, Buffered
Boiling Point 100°C
CAS 50-00-0, 67-56-1, 7732-18-5, 7558-79-4
Packaging Poly Bottle
Color Colorless
pH 6.9 to 7.1
Quantity 4 L
MDL Number MFCD00003274
Percent Purity 4.0 to 4.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.