Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP, 35 wt. %, Honeywell™ Riedel-de Haen™

Catalog No. 60044882 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
60 kg
6 x 1 L
Packaging:
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-882 4 x 2.5 L Poly Bottle
60-044-881 2.5 L Poly Bottle
60-044-884 6 x 1 L Poly Bottle
60-044-883 60 kg Poly Pail
60-044-880 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-044-882 Supplier Honeywell Chemicals Supplier No. 155134X2.5L

Meets analytical specification of BP, Ph. Eur., 35 wt. %, contains 10% methanol as stabilizer

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
SMILES C=O

Specifications

Melting Point <-15°C
Color Colorless
Density 1.09g/mL at 25°C
pH 3.5 to 4.5 (at 20°C)
Boiling Point 93°C to 96°C
Flash Point 64°C (147.2°F)
Ignition Residue ≤0.01% (as SO4)
Packaging Poly Bottle
Linear Formula HCHO
Quantity 4 x 2.5 L
UN Number UN2209
Beilstein 1209228
IUPAC Name formaldehyde
Formula Weight 30.03g/mol
Percent Purity 35 wt. %
Chemical Name or Material Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP
Show More Show Less
Hazard Statement P201-P260-P280-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P308 + P311
Precautionary Statement H301 + H311 + H331-H314-H317-H335-H341-H350-H370
DOTInformation 8/PGIII
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.