Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formaldehyde, 10% (v/v), Buffered, pH 7.0, Histological Fixative, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
250 mL
500 mL
1 L
4 L
10 L
20 L
Specifications
Specifications
| Packaging Type | Amber Glass Bottle |
| Concentration | 10% (v/v) |
| For Use With (Equipment) | All Tissue Processors |
| Grade | Chemical Solution |
| Product Type | Formaldehyde |
| Quantity | 250 mL |
| Solution Type | Formaldehyde |
| Form | Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction