Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 37%, Solution, Spectrum™ Chemical
SDP

Catalog No. 18602829
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum

F3328, 50-00-0, HCHO

Spectrum™ Chemical Formaldehyde, 37 Percent, Solution is also known as methanal and is the simplest aldehyde. It has many uses, one of which is formol, the famous embalming and tissue preserving liquid. It is also used in industrial application for the production of different types of resins. It is also a well known tissue fixative and was formerly a disinfectant.

Chemical Identifiers

CAS 50-00-0, 7732-18-5
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
IUPAC Name formaldehyde
SMILES C=O

Specifications

CAS Max % 37%, 50%
Packaging Poly Bottle
Quantity 1 L
UN Number UN1198
Percent Purity 34.5%
Grade Reagent
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.