Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formaldehyde, ACS, 36.5 to 38.0%, MilliporeSigma™
Shop All MilliporeSigma Products
Click to view available options
Packaging:
Plastic Bottle
Specifications
Specifications
Assay Percent Range | 36.5 to 38% |
Packaging | Plastic Bottle |
Quantity | 3.78 L |
Formula Weight | 30.03 |
Grade | ACS |
Physical Form | Solution |
Safety and Handling
Recommended Storage | Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction