Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde Dimethylhydrazone 98.0+%, TCI America™
SDP

Catalog No. F07131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07131G 1 g
F07135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07131G Supplier TCI America Supplier No. F07131G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2035-89-4
Molecular Formula C3H8N2
Molecular Weight (g/mol) 72.111
MDL Number MFCD01753011
InChI Key NDNVSJIXYFNRDR-UHFFFAOYSA-N
PubChem CID 74857
IUPAC Name N-methyl-N-(methylideneamino)methanamine
SMILES CN(C)N=C

Specifications

Melting Point -103°C
Color Colorless-Slightly Pale Yellow at 20°C
Boiling Point 71°C
Quantity 1 g
UN Number 1993
Formula Weight 72.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Formaldehyde Dimethylhydrazone
RTECSNumber LP9000000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.