Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Formaldehyde Fluorescent Detection Kit

Catalog No. eiach2o
Encompass_Preferred
Click to view available options
Quantity:
192 Tests

Includes

96-well plates, standard, other assay specific reagents, buffers and lot-specific technical data sheet.

Assay Formaldehyde levels in Human Urine and Tissue Culture Media.

The Formaldehyde Detection research-use-only kit is a fluorescent assay designed for the quantification and detection of formaldehyde in human urine and tissue culture media samples.

This complete, ready-to-use kit includes clear 96-well plate(s), formaldehyde standard, formaldehyde detection reagent, and other components to perform the assay. A 96-well fluorescence plate reader capable of reading fluorescent emission at 510 nm, with excitation at 450 nm, is required for use of this kit.

Performance characteristics
• Assay type: fluorescent detection Kit
• Sample types: human urine and tissue culture media samples
• Sensitivity: 0.715 μM
• Standard curve range: 3.1–200 μM
• Reactivity: species independent

Background
Formaldehyde is widely employed in industry for a wide range of applications. It acts as a disinfectant and biocide, and is utilized as a tissue fixative and embalming agent. Formaldehyde is a key metabolite produced during normal metabolism and contributes to methylation reactions and protein and nucleic acid synthesis. This kit was shown to measure formaldehyde in human urine and tissue culture media. Formaldehyde is identical across all species and cell types.

Assay principle
The Formaldehyde Urinary Detection kit designed to quantitatively measure formaldehyde present in tissue culture media and urine samples. A formaldehyde standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a black microtiter plate. The fluorescent reaction is initiated with the formaldehyde detection reagent, which is pipetted into each well. After a short incubation the emission of the generated fluorescent signal is detected in a microtiter plate reader capable of measuring 510 nm fluorescence utilizing 450 nm excitation wavelength.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 96-well plate(s), standard, other assay specific reagents, buffers, and lot-specific technical data sheet.

Store at –20°C.
Immunoassay Kit Format Fluorescent Detection Kit
Product Type Formaldehyde Fluorescent Detection Kit
Sample Type Urine
For Use With (Equipment) Microplate Reader
Gene ID (Entrez) Non-Gene
Gene Symbol Non-Gene
Quantity 192 Tests
Shipping Condition Approved for shipment on Wet or Dry Ice
Target Kit Named Formaldehyde

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.