Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, Para (Certified), Fisher Chemical

Catalog No. o4042500 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy

Catalog No. O4042-500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Chemical Identifiers

CAS 30525-89-4, 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point 120°C
Color White
Density 1.46g/cm┬│
pH 3.5 to 5.0
Assay Percent Range ≥95 %
Packaging Poly Bottle
Quantity 500 g
UN Number 2213
Vapor Pressure 1.2mmHg at 25┬░C
Percent Purity ≥95%
Grade Certified
Physical Form Solid
Chemical Name or Material Formaldehyde, Para
Show More Show Less
Hazard Category Signal Word: DANGER!
Precautionary Statement Emergency Overview
Flammable solid. Harmful by inhalation and if swallowed. Irritating to eyes, respiratory system and skin. Possible cancer hazard. May cause cancer based on animal data. May cause pulmonary edema.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.