Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP, 35 wt. %, Honeywell™ Riedel-de Haen™

Catalog No. 60044883
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
60 kg
6 x 1 L
Packaging:
Poly Bottle
Poly Pail

Meets analytical specification of BP, Ph. Eur., 35 wt. %, contains 10% methanol as stabilizer

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
SMILES C=O

Specifications

Melting Point <-15°C
Color Colorless
Density 1.09g/mL at 25°C
pH 3.5 to 4.5 (at 20°C)
Boiling Point 93°C to 96°C
Flash Point 64°C (147.2°F)
Ignition Residue ≤0.01% (as SO4)
Packaging Poly Pail
Linear Formula HCHO
Quantity 60 kg
UN Number UN2209
Beilstein 1209228
IUPAC Name formaldehyde
Formula Weight 30.03g/mol
Percent Purity 35 wt. %
Chemical Name or Material Formaldehyde Solution, Meets analytical specification of Ph.Eur.BP
Show More Show Less
Hazard Statement P201-P260-P280-P301 + P310 + P330-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P308 + P311
Precautionary Statement H301 + H311 + H331-H314-H317-H335-H341-H350-H370
DOTInformation 8/PGIII
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.