Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formalin, 10%, MilliporeSigma™
Shop All MilliporeSigma Products
Click to view available options
Quantity:
20 L
Specifications
Specifications
| Physical Form | Liquid |
| Color | Colorless |
| Quantity | 20 L |
| Assay Percent Range | 10% |
| Chemical Name or Material | Formalin, 10%, Fixative |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction