Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formalin 10% Neutral Buffered, Vion Biosciences
SCI_ED

Catalog No. p-200076282 Shop All Aldon Products
Click to view available options
Quantity:
500 mL
1 L
5 gal.
4 x 1 gal.
WARNING: This product can expose you to chemicals including Methanol and Formaldehyde, which are known to the State of California to cause cancer and birth defects or other reproductive harm.
WARNING: This product can expose you to chemicals including Methanol and Formaldehyde, which are known to the State of California to cause cancer and birth defects or other reproductive harm.

This 10% neutral buffered formalin is a stable, neutral fixative used to ensure the long-term preservation of tissues as close to the living state as possible and provide clear, reproducible results.

  • This 10% neutral buffered formalin is a stable, neutral fixative used to ensure the long-term preservation of tissues as close to the living state as possible and provide clear, reproducible results.

Specifications

Boiling Point ~100°C (212°F in water)
Closure Type Screw Cap
Color Colorless
Content And Storage Store in a cool, well-ventilated area away from incompatible substances.
Packaging Type HDPE Bottle
Concentration 10%
For Use With (Equipment) Microscope
Grade Histological
Product Line Vion Histology
Product Type Fixative
Quantity 500 mL
Solution Type Formalin
Formulation 10% Neutral Buffered Formalin
For Use With (Application) Histology
pH 7
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.