Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formalin, Buffered, 10% (Acetate Buffer/Certified), Fisher Chemical™

Catalog No. p-213365 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
20 L
Packaging:
Poly Bottle
PolyPac™

Specifications

Physical Form Liquid
Chemical Name or Material Formalin, Buffered, 10%
Melting Point 0°C
Name Note Acetate Buffer
CAS 50-00-0, 67-56-1, 127-09-3, 7732-18-5
Density 1.10g/cm³
Packaging Poly Bottle
pH 7
Quantity 4 L
MDL Number MFCD00003274
Percent Purity 4.0 to 5.0%
Grade Certified
Concentration or Composition (by Analyte or Components) 10% w/v (Acetate Buffer)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.