Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Bullfrogs
SCI_ED

For use in introducing students to vertebrate anatomy. Carolina™ Formalin Bullfrogs are ideal for studying amphibian anatomy, adaptations, and body systems.

$17.00

Specifications

Preservative Formalin
Length (English) 5+ in.
Length (Metric) 12.7+ cm
Product Type Preserved specimen
View More Specs

Products 3
Catalog Number Mfr. No. Injection Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Injection Type Quantity Price Quantity & Availability  
S37638
SCI_ED
View Documents
Carolina™
S227101
Plain 1/bag
Each for $17.00
Case of 25 EA for $418.80
Add to cart
 
S37707
SCI_ED
View Documents
Carolina™
S227102
Plain 5/bag This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S37639
SCI_ED
View Documents
Carolina™
S227121
Double 1/bag This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde
  • INJECTIONS: Plain: no color injection; Double: red arteries, blue hepatic
Specifications

Specifications

Formalin
5+ in.
12.7+ cm
Preserved specimen
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit