Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carolina™ Formalin Cats

Shop All Carolina Biological Supply Products


Click to view available options
Injection Type:
Double
Plain
Single
Triple
Length (English):
14 to 18 in.
18+ in.
Description
- Felis catus
- Similar to human anatomy
- Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
- Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde
- INJECTIONS: Plain: n
Specifications
Specifications
| Type | Preserved Specimen |
| Preservative | Formalin |
| Injection Type | Single |
| Length (English) | 18+ in. |
| Length (Metric) | 45.7+ cm |
| Quantity | 1/Bag |
| Packaging | Bag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction