Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Non-Venomous Snake
SDPSCI_ED

Catalog No. s37648 Shop All Carolina Biological Supply Products
Click to view available options
Quantity:
1/Bag

For use in learning more about reptiles by investigating anatomy of nonvenomous snake. Carolina™ Formalin Non-Venomous Snake is ideal for comparative vertebrate courses.

  • Species vary based on availability
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Length (English) 18 to 36 in.
Length (Metric) 45.7 to 91.4 cm
Quantity 1/Bag
Packaging Bag
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.