Learn More
Carolina™ Formalin Preserved Sheep Brain, Dura Mater Removed
SDPSCI_ED
Catalog No. p-7226621 Shop All Carolina Biological Supply Products
Click to view available options
Packaging:
Bag
Pail
This item is not returnable. View return policy
This item is not returnable. View return policy
Carolina™ Formalin Preserved Sheep Brain with the Dura Mater Removed is used to study mammalian brain structure.
  • Similar to human brain
  • Sheep brain with dura mater removed and optic chiasma intact are shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Odorless fluid designed to minimize unpleasant odor of formaldehyde
  • Plain: No color injection
  • Individually damp-packed in vacuum-sealed bags
Specifications
Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Quantity 1/Pk.
Packaging Pail
Autoclavable Not Autoclavable
Sterility Non-sterile
Reusable No
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.