Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Preserved Sheep Eyes
SDPSCI_ED

Catalog No. s28068 Shop All Carolina Biological Supply Products
Click to view available options
Quantity:
10/Pk.

Explores eye structures representative of mammals.

  • Packaged in vacuum-sealed bag
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe™ is an odorless fluid designed to minimize the unpleasant odor of formaldehyde

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Quantity 10/Pk.
Packaging Bulk Bag
Autoclavable Not Autoclavable
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.