Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Sea Anemones
SDPSCI_ED

Catalog No. S37685 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Size:
Base: 1.5 to 2 in.
Base: 2+ in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S37685 Base: 2+ in.
S37686 Base: 1.5 to 2 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S37685 Supplier Carolina™ Supplier No. 224228
This item is not returnable. View return policy

Carolina™ Formalin Sea Anemones belongs to Metridium sp.

  • Phylum Cnidaria; Class Anthozoa
  • Sessile, marine polyp
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
  • Carosafe is odorless fluid designed to minimize unpleasant odor of formaldehyde

Specifications

Type Preserved Specimen
Preservative Formalin
Injection Type Plain
Length (English) 2 in.
Packaging Jar
Autoclavable Not Autoclavable
Size Base: 2+ in.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.