Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Formalin Solution, 10%
SCI_ED

Catalog No. S25330 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
10% w/v
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25330 4 L 10% w/v
S25689 1 L 10% w/v
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25330
This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 50-00-0 , 67-56-1 , 7732-18-5
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Liquid
Packaging 10% w/v
Color Colorless
Quantity 4 L
Linear Formula HCHO
Grade Laboratory
Chemical Name or Material Formalin Solution, 10%
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 9, III, : Miscellaneous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.