Alfa Aesar™ Formamide, ACS, 99.5+%

Manufacturer: Alfa Aesar™ 1483530

Quantity 250g

 View more versions of this product

Catalog No. AA1483530

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 75-12-7
    Molecular Formula HCONH2
    Linear Formula HCONH2
    Formula Weight 45.04
    MDL Number MFCD00007941
    Beilstein 505995
    Merck Index 14,4237
    Chemical Name or Material Formamide, For ACS analysis
    Grade ACS Grade

    Specifications

    Formula Weight 45.04
    Assay Percent Range >99.5%
    Molecular Formula HCONH2
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Ammonia-like
    Health Hazard 2 GHS H Statement
    H360
    May damage fertility or the unborn child.
    Refractive Index 1.4475
    Melting Point 2°C
    Sensitivity Hygroscopic
    Merck Index 14,4237
    RTECS Number LQ0525000
    Boiling Point 210°C
    Physical Form Liquid
    Quantity 250g
    MDL Number MFCD00007941
    CAS 75-12-7
    Flash Point 154° (310°F)
    Density 1.134g/mL
    Color Colorless
    Beilstein 505995
    TSCA TSCA
    Linear Formula HCONH2
    Health Hazard 1 Danger
    Chemical Name or Material Formamide, For ACS analysis
    Assay ≥99.5%
    Grade ACS Grade

    GHS H Statement
    H360
    May damage fertility or the unborn child.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit