Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Formamide, Deionized, Ultrapure, 99.5%, Spectrum™ Chemical

Click to view available options
Quantity:
500 mL
950 mL
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Formamide, Deionized, Ultrapure - Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates. It is used as a RNA stabilizer in gel electrophoresis by deionizing RNA, in capillary electrophoresis to stabilize (single) strands of denatured DNA and as a constituent of cryoprotectant vitrification mixtures used for cryopreservation of tissues and organs.
Specifications
Specifications
CAS Max % | 100% |
Packaging | Amber Glass Bottle |
Quantity | 950 mL |
Percent Purity | 99.5% |
Grade | Ultrapure |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction