Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formamide, Ultra Pure, 99.5% min

Catalog No. p-7057714
Click to view available options
Quantity:
100 mL
250 mL

CAS: 75-12-7 | CH3NO | 45.04 g/mol

Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamide is used in the synthesis of medicine, animal glue and other organic compound. It is also used as a solvent which ionizes water-insoluble compounds. It is used as a softening agent for paper and fiber. It is widely used in the fields of the spice, dye, plastic as well as textile industries.

Solubility
Soluble in water (20 mg/ml ), methanol, ethanol and acetone. Slightly soluble in ether.

Notes
Hygroscopic. Store at room temperature. Store away from water or moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-12-7
Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
PubChem CID 713
ChEBI CHEBI:48431
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2°C
Color Colorless
Density 1.134 g/mL
Boiling Point 210°C
Quantity 100 mL
Flash Point 150°C (302°F)
Odor Ammonia-like
Physical Form Liquid
Refractive Index 1.4475
Beilstein 505995
Solubility Information Soluble in water (20mg/ml ),methanol,ethanol and acetone. Slightly soluble in ether.
Formula Weight 45.04
Percent Purity ≥99.5%
Chemical Name or Material Formamide, Ultra Pure
Show More Show Less
Hazard Category H351-H360FD-H373
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P260-P281-P308+P313-P501c
EINECSNumber 200-842-0
RTECSNumber LQ0525000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.